Learn More
Chemglass Life Sciences #24 Loosening Ring
SDP
Catalog No. 501531933
Encompass_Preferred
#24 Loosening Ring. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.
Catalog No. 50-153-1933 Supplier Chemglass Life Sciences Supplier No. CGB18403
#24 Loosening Ring. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.