Learn More
Stellar Scientific 24W, POLY DW PL NON-ST, 50/CS
SDP
Catalog No. NC2290983
Encompass_Preferred
24 Well Polypropylene Deep Well Plate, U-Bottom, Square Well, Rnase and Dnase Free, Non-Sterile, 5/PK, 50/CS
This item is not returnable. View return policy
Catalog No. NC2290983 Supplier Stellar Scientific Supplier No. ILPDP5MUS24N
24 Well Polypropylene Deep Well Plate, U-Bottom, Square Well, Rnase and Dnase Free, Non-Sterile, 5/PK, 50/CS