Learn More
Cell Biolabs Inc 24W WOUND HEALING TRIAL SIZE
Catalog No. 50672649
Encompass_Preferred
The CytoSelect 24-Well Wound Healing Assay (Trial Size) provides more consistent results well-to-well compared to homemade scratch assays. Useful for measuring cell migration, cell proliferation and wound closure.
Catalog No. 50-672-649 Supplier Cell Biolabs Inc Supplier No. CBA120T
The CytoSelect 24-Well Wound Healing Assay (Trial Size) provides more consistent results well-to-well compared to homemade scratch assays. Useful for measuring cell migration, cell proliferation and wound closure.