Learn More
Chemglass Life Sciences 24 Well Cell Culture Plate, Flat, TC, Sterile, Bulk, 10/pk, 50/cs
SDP
Catalog No. 501897677
Encompass_Preferred
Nest cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform growth. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.Made from high clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityBulk packaging - 10 plates per bagNon-Pyrogenic, DNase/RNase-Free
Catalog No. 50-189-7677 Supplier Chemglass Life Sciences Supplier No. CGN3500B24
Add to Cart
Add to Cart
Nest cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform growth. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.Made from high clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityBulk packaging - 10 plates per bagNon-Pyrogenic, DNase/RNase-Free