Learn More
CELLTREAT Scientific Products 24 Well Tissue Culture Plate with Lid, 5/Pack, Sterile
SDP
Catalog No. NC3002526
Encompass_Preferred
Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachmentPolystyrene is optically clear for easy cell viewing and imagingEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationPlates are labeled with alpha-numeric markings for easy identification of individual wellsLids have a stacking feature for added stability when handling tall stacks of platesRound bottom option maximizes sample retrieval and allows mixing (PN 229190)Plate is printed with lot number and expiration date for easy identification and trackingSturdy individual blister pack prevents damage when handling or shippingBulk packaging option (5/sleeve) increases efficiency for high volume users (PNs 229197 & 229127)
Catalog No. NC3002526 Supplier CELLTREAT Scientific Products Supplier No. 229127
Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachmentPolystyrene is optically clear for easy cell viewing and imagingEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationPlates are labeled with alpha-numeric markings for easy identification of individual wellsLids have a stacking feature for added stability when handling tall stacks of platesRound bottom option maximizes sample retrieval and allows mixing (PN 229190)Plate is printed with lot number and expiration date for easy identification and trackingSturdy individual blister pack prevents damage when handling or shippingBulk packaging option (5/sleeve) increases efficiency for high volume users (PNs 229197 & 229127)