Learn More
Caplugs 24WELL PLTS W/LID STERILE UNT
Catalog No. NC2459880
Encompass_Preferred
Caplugs Evergreen 24-Well Plates are available non-sterile, sterile, and treated for tissue culture. Sterile and treated plates are gamma-irradiated for sterility and are individually packed with lids.Distinct alpha-numeric well designationsWell volume: 3.5 mLLid sold separately6-well, 12-well, 96-well and 384-well units are also available - Items are not available for shipment to Mexico
Catalog No. NC2459880 Supplier Caplugs Supplier No. 222804401F
Caplugs Evergreen 24-Well Plates are available non-sterile, sterile, and treated for tissue culture. Sterile and treated plates are gamma-irradiated for sterility and are individually packed with lids.Distinct alpha-numeric well designationsWell volume: 3.5 mLLid sold separately6-well, 12-well, 96-well and 384-well units are also available - Items are not available for shipment to Mexico