Learn More
Gawet Marble and Granite Vermont Marble™ Slab, 24″ x 24″ x 3″ thick
Catalog No. NC0316472
Encompass_Preferred
The Vermont Marble™ Slab is for use on laboratory furniture.   It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads
Catalog No. NC0316472 Supplier Gawet Marble and Granite Supplier No. 24X24X3
The Vermont Marble™ Slab is for use on laboratory furniture.   It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads