Learn More
Uline Gas Cylinder Wall Bracket - 2 Cylinder Capacity | | 1 EA
Supplier:  Uline H10677
Encompass
Easy-to-use tank storage, Polyester straps with steel buckles hold tanks in place, Cushioned bumper guards for scratchless grip, Fits 4-12 diameter cylinders, Meets NFPA and OSHA Standards, Mounting hardware not included
Catalog No. NC2750538