Learn More
Tewes Corporation 24X30 4MIL FLAT BAG 250/CS
SDP
Catalog No. NC0239534
Encompass_Preferred
Flat Poly Bags with Heavy Duty Protection when Packaging heavier weight or bulkier items; Strong enough to reuse multiple times; Meet FDA and USDA specifications; Bags can be heat sealed, tied,,stapled or taped to close. 250/CASE.
Catalog No. NC0239534 Supplier Tewes Corporation Supplier No. F24304
Flat Poly Bags with Heavy Duty Protection when Packaging heavier weight or bulkier items; Strong enough to reuse multiple times; Meet FDA and USDA specifications; Bags can be heat sealed, tied,,stapled or taped to close. 250/CASE.