Learn More
Essentra Porous Technologies FG Clean Wipes, High Absorbency, Lint-Free, 24 x 36 in
SDP
Catalog No. NC1856195
Encompass_Preferred
FG CLEAN WIPES are high-performance nonwoven polyester and cellulose cleanroom wipers designed for critical environments. With superior absorbency and low linting, they ensure effective spill cleanup and surface decontamination, minimizing contamination risks. Engineered free of binders and additives, they are compatible with various cleaning agents. Ideal for ISO 5-8 environments, each case contains 180 lint-free wipes, making them a reliable choice for maintaining safety and cleanliness in sensitive scientific settings.
Catalog No. NC1856195 Supplier Essentra Porous Technologies Supplier No. 7C1ABGRPSP00
FG CLEAN WIPES are high-performance nonwoven polyester and cellulose cleanroom wipers designed for critical environments. With superior absorbency and low linting, they ensure effective spill cleanup and surface decontamination, minimizing contamination risks. Engineered free of binders and additives, they are compatible with various cleaning agents. Ideal for ISO 5-8 environments, each case contains 180 lint-free wipes, making them a reliable choice for maintaining safety and cleanliness in sensitive scientific settings.