Learn More
Ace Glass, Inc. 24X50 SIL/MET CLAMP (6PK)
SDP
Supplier:  Ace Glass, Inc. 759925
Encompass
24/40 nickel coated steel clamp to snap on/off standard taper joints with ease while not scratching the glass. Clip offers superior chemical and temperature resistance (500° C).
Catalog No. NC9002813