Learn More
HIMEDIA Durham Tubes Neutral glass, autoclavable;
length : 25mm - 27mm, 
diameter : 6 - 7mm.
Catalog No. NC1205535
Encompass_Preferred
Durham Tubes
Catalog No. NC1205535 Supplier HIMEDIA Supplier No. GW1631X100NO
Durham Tubes