Learn More
Leica Biosystems Set of specimen discs
Catalog No. NC0558768
Encompass_Preferred
Set of specimen discs used as cryostat chucks for mounting tissue specimens during cryosectioning. Leica Biosystems lists this item under manufacturer part number 14047043550; the official product page does not specify diameter or pack quantity, so confirm those details before ordering.
  • Provide a stable surface for mounting tissue specimens for cryostat sectioning.
  • Compatible with common cryostat models.
  • Sold as a set; verify diameter and pack quantity on order.
  • Designed to hold specimens securely during microtomy.
Catalog No. NC0558768 Supplier Leica Biosystems Supplier No. 14047043550
Set of specimen discs used as cryostat chucks for mounting tissue specimens during cryosectioning. Leica Biosystems lists this item under manufacturer part number 14047043550; the official product page does not specify diameter or pack quantity, so confirm those details before ordering.
  • Provide a stable surface for mounting tissue specimens for cryostat sectioning.
  • Compatible with common cryostat models.
  • Sold as a set; verify diameter and pack quantity on order.
  • Designed to hold specimens securely during microtomy.