Learn More
Thermo Scientific Thermo Scientific Tygon Tubing 25.4mm O.D.
Supplier:  Thermo Scientific 003010100020
Encompass
25.4 millimeter diameter flexible chemical-resistant tubing often used for fluid transfer in lab equipment has 0.984 inch inner diameter, sold by the foot, withstands temperatures from -60 to 150 degrees Celsius with broad chemical compatibility and minimal leaching and absorption.
Catalog No. NC0273067