Learn More
Chemglass Life Sciences 25.4mm PTFE Gasket
SDP
Catalog No. CG204903
Encompass_Preferred
25.4mm PTFE Gasket. PTFE gasket is 0.015" thick with an opening to fit Chemglass stirrer shafts. Gasket is placed below the CG-2048 Safety collar to prevent wear as a result of the collar rotating against the stirrer bearing. Also useful as a dust cover. SOLD 12/PKG.
Catalog No. CG204903 Supplier Chemglass Life Sciences Supplier No. CG204903
25.4mm PTFE Gasket. PTFE gasket is 0.015" thick with an opening to fit Chemglass stirrer shafts. Gasket is placed below the CG-2048 Safety collar to prevent wear as a result of the collar rotating against the stirrer bearing. Also useful as a dust cover. SOLD 12/PKG.