Learn More
Chemglass Life Sciences Chemglass 25.4 mm Rotary Bearing Rebuild Kit for Mechanical Seal Stir Bearings (CG-2077-M-325)
SDP
Supplier:  Chemglass Life Sciences CG2077M325
Encompass_Preferred
The CG-2077-M-325 25.4 mm rotary bearing rebuild kit provides replacement sealing components to restore mechanical seal stir bearings to vacuum- and pressure-tight operation. The kit includes perfluorinated O-rings and related parts, and is supplied with a gold-plated wrench for installation and adjustment.
  • Restores rotary bearing seals to maintain vacuum and pressure tight operation.
  • Contains perfluorinated O-rings and replacement sealing components.
  • Designed for 25.4 mm shaft and bearing applications.
  • Compatible with mechanical seal stir bearing assemblies.
  • Supplied with a gold-plated wrench for installation.
  • Helps extend seal life and reduce contamination from flaking.
  • Supports vacuum up to 5 Torr and pressure up to 2-3 PSI.
Catalog No. 50-273-3534