Learn More
Chemglass Life Sciences Bearing Body Only, 25.4mm Stir Bearing, Mechanical Seal, 2-3/8in Threaded Connection
SDP
Catalog No. 50872510
Encompass_Preferred
Bearing Body Only, 25.4mm Stir Bearing, Mechanical Seal, 2-3/8in Threaded Connection. Our NEW mechanical seal bearing is the only design on the market that will permit you to use your current glass, metal or PTFE covered stir shaft. The design allows you to have the flexibility of vertical adjustment and still choose your agitator diameter and style. Bearing has a ferrule and locking nut to ensure that the seal rotates properly, and is supplied complete with a gold plated wrench. Lower joint adapters are supplied complete with perfluorinated o-rings and a loosening nut. PLEASE NOTE: BEARING BODY AND LOWER JOINT ADAPTER MUST BE ORDERED SEPARATELY.**MAX RPM WILL DEPEND ON SHAFT AND/OR AGITATOR TYPE, ALONG WITH ALIGNMENT OF MOTOR AND COUPLING. CHEMGLASS RECOMMENDS A MAX OF 500 RPM WHEN USED WITH ANY GLASS VESSEL, LID OR SHAFT. LONGER LENGTH STIRRER SHAFTS ARE REQUIRED FOR USE WITH CG-2077-M MECHANICAL SEAL STIR BEARINGS.
Catalog No. 50-872-510 Supplier Chemglass Life Sciences Supplier No. CG2077M04
Bearing Body Only, 25.4mm Stir Bearing, Mechanical Seal, 2-3/8in Threaded Connection. Our NEW mechanical seal bearing is the only design on the market that will permit you to use your current glass, metal or PTFE covered stir shaft. The design allows you to have the flexibility of vertical adjustment and still choose your agitator diameter and style. Bearing has a ferrule and locking nut to ensure that the seal rotates properly, and is supplied complete with a gold plated wrench. Lower joint adapters are supplied complete with perfluorinated o-rings and a loosening nut. PLEASE NOTE: BEARING BODY AND LOWER JOINT ADAPTER MUST BE ORDERED SEPARATELY.**MAX RPM WILL DEPEND ON SHAFT AND/OR AGITATOR TYPE, ALONG WITH ALIGNMENT OF MOTOR AND COUPLING. CHEMGLASS RECOMMENDS A MAX OF 500 RPM WHEN USED WITH ANY GLASS VESSEL, LID OR SHAFT. LONGER LENGTH STIRRER SHAFTS ARE REQUIRED FOR USE WITH CG-2077-M MECHANICAL SEAL STIR BEARINGS.