Learn More
Chemglass Life Sciences Optional 25.4mm Stirrer Shaft, Reactor Size: 50L (Tall), Fits Support Frame: CG-1968-F-14, Overall Length: 48.2"
SDP
Catalog No. CG209752
Encompass_Preferred
25.4mm Stirrer Shaft, Reactor Size: 50L (Tall), Fits Support Frame: CG-1968-F-14, Overall Length: 48.2". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. CG209752 Supplier Chemglass Life Sciences Supplier No. CG209752
25.4mm Stirrer Shaft, Reactor Size: 50L (Tall), Fits Support Frame: CG-1968-F-14, Overall Length: 48.2". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.