Learn More
Chemglass Life Sciences Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L (Short), Fits Support Frame: CG-1968-X-12, Overall Length: 44.9"
SDP
Catalog No. 50872528
Encompass_Preferred
Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L (Short), Fits Support Frame: CG-1968-X-12, Overall Length: 44.9". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat.REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. 50-872-528 Supplier Chemglass Life Sciences Supplier No. CG209751
Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L (Short), Fits Support Frame: CG-1968-X-12, Overall Length: 44.9". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat.REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.