Learn More
Chemglass Life Sciences Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L (Tall), Fits Support Frame: CG-1968-X-14, Overall Length: 51.7"
SDP
Catalog No. 50872529
Encompass_Preferred
Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L (Tall), Fits Support Frame: CG-1968-X-14, Overall Length: 51.7". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat.REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. 50-872-529 Supplier Chemglass Life Sciences Supplier No. CG209753
Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L (Tall), Fits Support Frame: CG-1968-X-14, Overall Length: 51.7". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat.REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.