Learn More
Lonza Walkersville Tissue Culture Plates, 25 Count
Catalog No. NC9014111
Encompass
Clear Bottom White Walled Tissue Plates are white walled 96-well plastic plates designed specifically for use with any bioluminescent bioassay kit.Content:25 x Clear Bottom, White-Walled Tissue Culture Plates
Catalog No. NC9014111 Supplier Lonza Walkersville Supplier No. LT27102
Clear Bottom White Walled Tissue Plates are white walled 96-well plastic plates designed specifically for use with any bioluminescent bioassay kit.Content:25 x Clear Bottom, White-Walled Tissue Culture Plates