Learn More
National Safety Apparel™ 25 Cal 32 in. Arc Flash Coat
SDP
Catalog No. 17001268 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-268 X-Large
17-001-261 Small
17-001-262 Medium
17-001-263 Large
17-001-264 2X-Large
17-001-265 3X-Large
17-001-266 4X-Large
17-001-267 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-268 Supplier National Safety Apparel™ Supplier No. C04UTUK25XL32
Coat provides the highest level of protection in work with electrical exposures and hazards
  • Breathable FR fabric
  • FR fabric will NOT melt, drip or ignite
  • Woven FR wrist cuffs
  • Generous fit for easy movement
  • Front hook and loop closure
  • Inherently flame resistant for FR Protection that will NOT wash off.
Specifications
Closure Type FR Hook and Loop
Color Khaki
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 25 cal/cm²
Certifications/Compliance CAT 3, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462