Learn More
Heska Corporation Element POC Test Card with BUN, Rapid Veterinary Diagnostics, Room Temperature Storage, Unit of Measure: 25 Count
SDP
Catalog No. NC2011212
Encompass
The Element POC Test Card with BUN is a rapid diagnostic tool for veterinary settings, delivering critical blood gas, electrolyte, and chemistry analyses in under 60 seconds. Engineered for use with the Element POC analyzer, it offers 18 parameters from a small sample volume. Its room-temperature stability and automatic calibration minimize waste, making it an efficient choice for timely patient assessments.
Catalog No. NC2011212 Supplier Heska Corporation Supplier No. CAT 5424
The Element POC Test Card with BUN is a rapid diagnostic tool for veterinary settings, delivering critical blood gas, electrolyte, and chemistry analyses in under 60 seconds. Engineered for use with the Element POC analyzer, it offers 18 parameters from a small sample volume. Its room-temperature stability and automatic calibration minimize waste, making it an efficient choice for timely patient assessments.