Learn More
Cayman Chemical 25-Desacetyl Rifampicin, Purity: ≥95%, CAS: 16783-99-6, 1 mg
SDP
Catalog No. 502912735
Encompass_Preferred
A major active metabolite of rifampicin; active against M. smegmatis (MIC99 = 2.66 µM)
Catalog No. 50-291-2735 Supplier Cayman Chemical Supplier No. 348171 MG
A major active metabolite of rifampicin; active against M. smegmatis (MIC99 = 2.66 µM)