Learn More
Sigma Aldrich Fine Chemicals Biosciences 25-Diphenyloxazole 99 suit100G
Catalog No. 502897405
Encompass
2 5-Diphenyloxazole (PPO) is an oxazole that can be used as a scintillator and a laser dye. It has fluorescence at a wavelength of 375 nm. It can be used as a wavelength shifter in liquid samples due to their high quantum yield and large strokes shift.
Catalog No. 50-289-7405 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D210404100G
Add to Cart
Add to Cart
2 5-Diphenyloxazole (PPO) is an oxazole that can be used as a scintillator and a laser dye. It has fluorescence at a wavelength of 375 nm. It can be used as a wavelength shifter in liquid samples due to their high quantum yield and large strokes shift.