Learn More
Caron Products And Services Refrigerated Incubator, 25 cu. ft., Temp, No RH
Catalog No. NC2334559
Encompass_Preferred
25 ft3 (708 L) Refrigerated Incubator offers precise temperature control (5C to 70C). Ideal for a wide range of applications. This reach-in unit delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
Catalog No. NC2334559 Supplier Caron Products And Services Supplier No. 7001251
Add to Cart
Add to Cart
Request a Quote
25 ft3 (708 L) Refrigerated Incubator offers precise temperature control (5C to 70C). Ideal for a wide range of applications. This reach-in unit delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.