Learn More
Nordson EFD 27 Gauge All Purpose Tips Clear 50/pk
SDP
Catalog No. NC0692645
Encompass
This item has a minimum qty of 3 per supplier requirements.
Made from burr-free, polished passivated stainless steel, Optimum general purpose dispense tips are ideal for handling a wide range of assembly fluids.These tips deliver precise, consistent deposits every time. Designed with our own unique polypropylene SafetyLok™ hubs that fit securely onto the barrel reservoir, general purpose tips prevent leakage and ensure drip-free dispensing.Each hub is designed with flush mating of the hub to the cutoff for smooth fluid flow and to prevent trapped air, which causes dripping and drooling. Burr-free passivated stainless steel tubing ensures consistent fluid deposits. Each hub is also manufactured and quality-tested to be free of molding flash bits of plastic that can block fluid flow and cause inconsistent deposits.
This item is not returnable. View return policy
Catalog No. NC0692645 Supplier Nordson EFD Supplier No. 7018395
This item has a minimum qty of 3 per supplier requirements.
Made from burr-free, polished passivated stainless steel, Optimum general purpose dispense tips are ideal for handling a wide range of assembly fluids.These tips deliver precise, consistent deposits every time. Designed with our own unique polypropylene SafetyLok™ hubs that fit securely onto the barrel reservoir, general purpose tips prevent leakage and ensure drip-free dispensing.Each hub is designed with flush mating of the hub to the cutoff for smooth fluid flow and to prevent trapped air, which causes dripping and drooling. Burr-free passivated stainless steel tubing ensures consistent fluid deposits. Each hub is also manufactured and quality-tested to be free of molding flash bits of plastic that can block fluid flow and cause inconsistent deposits.