Learn More
Braintree Scientific 25 GAUGE BLUNT NEEDLES 12PK
SDP
Catalog No. NC9250450
Encompass
Blunt tip needle, 25G-1 in, Stainless steel w/standard luer plastic hub, Use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, Blood sampling and catheter flushing, 1/DZ **Sold for research purposes only and not for human use**
Catalog No. NC9250450 Supplier Braintree Scientific Supplier No. NB25G
Blunt tip needle, 25G-1 in, Stainless steel w/standard luer plastic hub, Use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, Blood sampling and catheter flushing, 1/DZ **Sold for research purposes only and not for human use**