Learn More
Cayman Chemical 25-hydroxy Cholesterol, Purity: ≥98%, CAS: 2140-46-7, 10 mg
SDP
Catalog No. 502951617
Encompass_Preferred
An oxysterol; suppresses endogenous cholesterol synthesis by binding INSIG proteins and preventing SREBPs from being transported to the Golgi; inhibits IgA class switching induced by LPS and various cytokines in B cells (IC50 = ~50 nM); inhibits replication of SARS-CoV-2 in Vero cells (EC50 = 3.675 µM) and reduces increases in viral protein production in infected Vero cells when used prior to infection; reduces viral RNA loads in the lung and trachea in a mouse model of SARS-CoV-2 infection at 100 mg/kg per day; serum levels are increased in patients with SARS-CoV-2.
Catalog No. 50-295-1617 Supplier Cayman Chemical Supplier No. 1109710 MG
An oxysterol; suppresses endogenous cholesterol synthesis by binding INSIG proteins and preventing SREBPs from being transported to the Golgi; inhibits IgA class switching induced by LPS and various cytokines in B cells (IC50 = ~50 nM); inhibits replication of SARS-CoV-2 in Vero cells (EC50 = 3.675 µM) and reduces increases in viral protein production in infected Vero cells when used prior to infection; reduces viral RNA loads in the lung and trachea in a mouse model of SARS-CoV-2 infection at 100 mg/kg per day; serum levels are increased in patients with SARS-CoV-2.