Learn More
Cayman Chemical 25-hydroxy Cholesterol, 2140-46-7, 10 mg
SDP
Supplier:  Cayman Chemical 1109710
Encompass_Preferred
A side-chain substituted oxysterol derived from dietary cholesterol that inhibits the cleavage of SREBPs; has been implicated in a variety of metabolic events including cholesterol homeostasis and atherosclerosis as well as antitumor and immunomodulating activities
Catalog No. NC1197439