Learn More
Cayman Chemical 25-hydroxy Cholesterol, 2140-46-7, 25 mg
SDP
Supplier:  Cayman Chemical 1109725
Encompass_Preferred
Molecular Formula C27H46O2, Purity ≥98%, Formula Weight 402.7, Synonyms: cholest-5-ene-3β,25-diol; A side-chain substituted oxysterol derived from dietary cholesterol that inhibits the cleavage of SREBPs; has been implicated in a variety of metabolic events including cholesterol homeostasis and atherosclerosis as well as antitumor and immunomodulating activities.
Catalog No. NC0662686