Learn More
Cayman Chemical 25-hydroxy Vitamin D2, Purity: ≥98%, CAS: 21343-40-8, 1 mg
SDP
Catalog No. 502954328
Encompass_Preferred
A metabolite of vitamin D2; serum levels of 25-hydroxy vitamin D2 have been used as a marker of vitamin D status
Catalog No. 50-295-4328 Supplier Cayman Chemical Supplier No. 120781 MG
A metabolite of vitamin D2; serum levels of 25-hydroxy vitamin D2 have been used as a marker of vitamin D status