Learn More
Cayman Chemical 25-hydroxy VItamIn D2 500ug
SDP
Supplier:  Cayman Chemical 12078500 UG
Encompass_Preferred
A metabolite of vitamin D2; serum levels of 25-hydroxy vitamin D2 have been used as a marker of vitamin D status
Catalog No. 50-295-4185