Learn More
Cayman Chemical 25-hydroxy Vitamin D2-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 1262843-46-8, 1 mg
SDP
Catalog No. 502911448
Encompass_Preferred
An internal standard for the quantification of 25-hydroxy vitamin D2 by GC- or LC-MS
Catalog No. 50-291-1448 Supplier Cayman Chemical Supplier No. 348531 MG
An internal standard for the quantification of 25-hydroxy vitamin D2 by GC- or LC-MS