Learn More
Cayman Chemical 25-hydroxy VItamIn D3 10mg
SDP
Catalog No. 502894997
Encompass_Preferred
An intermediate in the pathway leading to the production of 1,25-dihydroxy vitamin D3; plasma or serum levels are commonly used as an indicator of an individual’s vitamin D status
Catalog No. 50-289-4997 Supplier Cayman Chemical Supplier No. 900068310 MG
Add to Cart
Add to Cart
An intermediate in the pathway leading to the production of 1,25-dihydroxy vitamin D3; plasma or serum levels are commonly used as an indicator of an individual’s vitamin D status