Learn More
Medchemexpress LLC HY-113134 10mg Medchemexpress, 25-Hydroxycholesterol CAS:2140-46-7 Purity:>98%
SDP
Catalog No. 501969229
Encompass_Preferred
Medchemexpress, HY-113134 10mg 25-Hydroxycholesterol CAS:2140-46-7 25-Hydroxycholesterol is a metabolite of cholesterol that is produced and secreted by macrophages in response to Toll-like receptor (TLR) activation. 25-hydroxycholesterol is a potent (EC50≈65 nM) and selective suppressor of IgA production by B cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9229 Supplier Medchemexpress LLC Supplier No. HY11313410MG
Medchemexpress, HY-113134 10mg 25-Hydroxycholesterol CAS:2140-46-7 25-Hydroxycholesterol is a metabolite of cholesterol that is produced and secreted by macrophages in response to Toll-like receptor (TLR) activation. 25-hydroxycholesterol is a potent (EC50≈65 nM) and selective suppressor of IgA production by B cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.