Learn More
Abcam 25-Hydro x ycholesterol, cholesterol metabolite, 5MG
Catalog No. 502704927
Encompass_Preferred
MW 402.65 g/mol. Cholesterol metabolite produced and secreted by macrophages in response to TLR activation. Regulates immune function and has a key role in the pathogenesis of atherosclerosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4927 Supplier Abcam Supplier No. AB1461455MG
MW 402.65 g/mol. Cholesterol metabolite produced and secreted by macrophages in response to TLR activation. Regulates immune function and has a key role in the pathogenesis of atherosclerosis.
The product is subject to the following: Abcam Restricted Use Statement