Learn More
Enzo Life Sciences 25-Hydroxyvitamin D3 (1mg). CAS: 19356-17-3
SDP
Supplier:  Enzo Life Sciences BMLDM1000001
Encompass_Preferred
Intermediate in the biosynthesis of 1α,25-dihydroxyvitamin D3. Major circulating metabolite of vitamin D3. Alternative name: Calcifediol, 25-Hydroxycholecalciferol. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0513