Learn More
Analytical Sales & Services 25µL GC Syringe for LEAP/Gerstel CTC PAL GC Autosamplers
Catalog No. 5000804777
Encompass
25µL GC Syringe 1702N for LEAP/Gerstel CTC PAL GC Autosamplers.
26s Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH
Catalog No. 50-008-04777 Supplier Analytical Sales & Services Supplier No. 42030434
25µL GC Syringe 1702N for LEAP/Gerstel CTC PAL GC Autosamplers.
26s Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH