Learn More
Avanti Polar Lipids 25 MG OF 18:1 PHOS ACID
Catalog No. NC9155022
Encompass
18: 1 Lyso PA, Lysophospholipids, Molecular Weight: 458.51, Molecular Formula: C 21 H 40 O7PNa, Purity: > 99%, Powder form, White Color, Not Flammable, Storage: -20 deg.C, 25mg
Catalog No. NC9155022 Supplier Avanti Polar Lipids Supplier No. 857130P25MG
18: 1 Lyso PA, Lysophospholipids, Molecular Weight: 458.51, Molecular Formula: C 21 H 40 O7PNa, Purity: > 99%, Powder form, White Color, Not Flammable, Storage: -20 deg.C, 25mg