Learn More
Ibidi USA micro-Insert 4 Well for Self-Insertion, 25 Culture-Inserts in 10 cm Transport Dish
SDP
Catalog No. 50305782
Encompass
The micro–Insert 4 Well is a biocompatible, silicone insert ideal for long term microscopy and stem cell cultivation. The micro–Insert has a special sticky bottom that prevents leaking on clean, flat, and dry surfaces, such as microscopy slides and dishes. Adherent or non–adherent cells placed into the small wells sink to the bottom and can be analyzed microscopically. The wells provide rectangular digital format (4:3) for covering the entire area by CCD cameras on microscopes. Tapered walls provide excellent optical access to cells located near the edges of the well. After cell attachment, the micro-Insert can be removed by using sterile tweezers without leaving a residue. 25 micro-Insert 4 Well in 10 cm transport dish, sterile, 25 inserts/box
This item is not returnable. View return policy
Catalog No. 50-305-782 Supplier Ibidi USA Supplier No. 80409
The micro–Insert 4 Well is a biocompatible, silicone insert ideal for long term microscopy and stem cell cultivation. The micro–Insert has a special sticky bottom that prevents leaking on clean, flat, and dry surfaces, such as microscopy slides and dishes. Adherent or non–adherent cells placed into the small wells sink to the bottom and can be analyzed microscopically. The wells provide rectangular digital format (4:3) for covering the entire area by CCD cameras on microscopes. Tapered walls provide excellent optical access to cells located near the edges of the well. After cell attachment, the micro-Insert can be removed by using sterile tweezers without leaving a residue. 25 micro-Insert 4 Well in 10 cm transport dish, sterile, 25 inserts/box