Learn More
Electron Microscopy Sciences Compact 25 Microscope Slide Box, Red
SDP
Catalog No. 50931093
Encompass_Preferred
This 25 capacity slide box is made from rigid polystyrene, with a removable lid.
Color: Red
Capacity: 25 slides
Catalog No. 50-931-093 Supplier Electron Microscopy Sciences Supplier No. 71455R
This 25 capacity slide box is made from rigid polystyrene, with a removable lid.
Color: Red
Capacity: 25 slides