Learn More
Integra Biosciences Corp 25 ml Divided Reservoir - Polystyrene | 30 individually wrapped
SDP
Catalog No. NC1685477
Encompass
SureFloTM array spreads reagent evenly across the reservoir and allows tips to sit on the bottom without aspirating air. This further reduces dead volume, saving you money on reagents and retaining more precious samples!
This item is not returnable. View return policy
Catalog No. NC1685477 Supplier Integra Biosciences Corp Supplier No. 4351
SureFloTM array spreads reagent evenly across the reservoir and allows tips to sit on the bottom without aspirating air. This further reduces dead volume, saving you money on reagents and retaining more precious samples!