Learn More
Aqua Solutions FerroVer Iron Reagent Powder Pillows, 25 mL, pk/1000
SDP
Catalog No. NC1525016
Encompass
For Iron determination by the 1,10 phenanthroline method, using FerroVer Iron Reagent. Hach Method 8008. USEPA approved for reporting wastewater analysis. Sample size: 25 mL. Pack of 1000 Powder Pillows To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. NC1525016 Supplier Aqua Solutions Supplier No. HA85428
For Iron determination by the 1,10 phenanthroline method, using FerroVer Iron Reagent. Hach Method 8008. USEPA approved for reporting wastewater analysis. Sample size: 25 mL. Pack of 1000 Powder Pillows To confirm accurate results, see appropriate standard solutions on the Consumables tab.