Learn More
Electron Microscopy Sciences Air Sampling 25 mm Asbestos TEM Cassette
SDP
Supplier:  Electron Microscopy Sciences 6523001
Encompass
This Universal Cassette and Cyclone Holder allows cassettes and cyclones to be attached onto a workers collar. Compatible with 37 mm and 25 mm cassettes, and aluminum cyclone manufactured by Zefon and SKC. Weighs only 73 g or 2.5 oz. Comes with 3' of tubing attached Weighs 2.5 oz. Comes with 3' of tubing attached.
Catalog No. 50-338-84