Learn More
Apexbio Technology LLC 25 mM dNTP Mixture 5x1ml
SDP
Catalog No. 5000063401
Encompass_Preferred
The 25 mM dNTP Mixture is an aqueous solution comprising equimolar concentrations (25 mM each) of four essential nucleotides (dATP dCTP dGTP and dTTP) These nucleoside triphosphates function as fundamental substrates during DNA synthesis and amplification serving as building blocks for DNA polymerases Designed with careful neutralization to pH 7 0 using NaOH this mixture allows precise control in molecular biology protocols Common experimental applications include PCR real-time PCR DNA sequencing complementary DNA (cDNA) synthesis via reverse transcription and site-directed mutagenesis The readiness balanced molar proportion and standardized pH permit optimal conditions for nucleotide incorporation reactions in research laboratories
Catalog No. 50-000-63401 Supplier Apexbio Technology LLC Supplier No. K10425
The 25 mM dNTP Mixture is an aqueous solution comprising equimolar concentrations (25 mM each) of four essential nucleotides (dATP dCTP dGTP and dTTP) These nucleoside triphosphates function as fundamental substrates during DNA synthesis and amplification serving as building blocks for DNA polymerases Designed with careful neutralization to pH 7 0 using NaOH this mixture allows precise control in molecular biology protocols Common experimental applications include PCR real-time PCR DNA sequencing complementary DNA (cDNA) synthesis via reverse transcription and site-directed mutagenesis The readiness balanced molar proportion and standardized pH permit optimal conditions for nucleotide incorporation reactions in research laboratories