Learn More
Chemglass Life Sciences Thermometer, Non-Mercury, 10/30 Joint, 25mm Immersion, -10°C to 250°C
SDP
Catalog No. CG3506N12
Encompass_Preferred
Thermometer, Non-Mercury, 10/30 Joint, 25mm Immersion, -10¿C to 250¿C. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. Thermometer has a white back, a 6 to 7mm outer diameter, and a standard taper 10/30 inner ground joint. All ranges have 1¿ subdivisions. Immersion is measured from the bottom of the joint to the bottom of the mercury bulb.
Catalog No. CG3506N12 Supplier Chemglass Life Sciences Supplier No. CG3506N12
Thermometer, Non-Mercury, 10/30 Joint, 25mm Immersion, -10¿C to 250¿C. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. Thermometer has a white back, a 6 to 7mm outer diameter, and a standard taper 10/30 inner ground joint. All ranges have 1¿ subdivisions. Immersion is measured from the bottom of the joint to the bottom of the mercury bulb.