Learn More
Medchemexpress LLC 25-NBD Cholesterol | 105539-27-3 | 99.0% | 564.76 | 1 MG
SDP
Catalog No. 5000451954
Encompass_Preferred
25-NBD Cholesterol is a fluorescently labeled form of cholesterol that facilitates the distinction between exogenous and endogenous cholesterol within bacterial cell membranes, emitting green fluorescence signals.
  • Fluorescently labeled form of cholesterol.
  • Facilitates distinction between exogenous and endogenous cholesterol within bacterial cell membranes.
  • Emits green fluorescence signals.
Catalog No. 50-004-51954 Supplier Medchemexpress LLC Supplier No. HYW0088841MG
Add to Cart
Add to Cart
25-NBD Cholesterol is a fluorescently labeled form of cholesterol that facilitates the distinction between exogenous and endogenous cholesterol within bacterial cell membranes, emitting green fluorescence signals.
  • Fluorescently labeled form of cholesterol.
  • Facilitates distinction between exogenous and endogenous cholesterol within bacterial cell membranes.
  • Emits green fluorescence signals.