Learn More
Enzo Life Sciences 25(OH) Vitamin D ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002150001
Encompass_Preferred
Rapid, easy-to-use ELISA kit to quantify 25(OH) Vitamin D for bone metabolism, nutrition, endocrinology, immunity and cancer. The 25(OH) Vitamin D ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 25(OH) Vitamin D in plasma and serum with results in just 1.5 hours. Alternative name: 25-Hydroxyvitamin D. Sensitivity: 1.98 ng/ml (range 0.5-1010 ng/ml). Use/Stability: All kit components should be stored at -20ºC. Assay Time: ~1.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human . Content: Microtiter Plate, Conjugate Concentrate, Antibody, Dissociation Buffer, Conjugate Diluent, Sample Diluent, Wash Buffer Concentrate, Standards, Substrate, Stop Solution, Assay Layout.
Catalog No. 50-200-6869