Learn More
MOLECULAR DEPOT LLC 25-OH Vitamin D3 Biotin - 1 mg
Catalog No. 5000013381
Encompass_Preferred
25-OH Vitamin D3 Biotin Conjugate consists of 25-OH Vitamin D3 conjugated to Biotin via a 22 A spacer arm The spacer adds flexibility to the 25-OH Vitamin D3 moiety which is required for its interaction with specific antibodies and other binding partners This compound has been tested for use in several 25-OH Vitamin D applications including ELISA The conjugate is supplied as a DMSO stock solution For in vitro research use only
Catalog No. 50-000-13381 Supplier MOLECULAR DEPOT LLC Supplier No. B2010192
25-OH Vitamin D3 Biotin Conjugate consists of 25-OH Vitamin D3 conjugated to Biotin via a 22 A spacer arm The spacer adds flexibility to the 25-OH Vitamin D3 moiety which is required for its interaction with specific antibodies and other binding partners This compound has been tested for use in several 25-OH Vitamin D applications including ELISA The conjugate is supplied as a DMSO stock solution For in vitro research use only